3-Hydroxypyrido[1,2-a]indole-10-carbonitrile ≥95%

Supplier: Matrix Scientific

045637-500MG 045637-1G
101871-874EA 375.38 USD
101871-874 101871-876
3-Hydroxypyrido[1,2-a]indole-10-carbonitrile ≥95%
3-Hydroxypyrido[1,2-a]indole-10-carbonitrile

Hazard: Irritant

Formula: C₁₃H₈N₂O
Melting Pt: 239…240 °C
MDL Number: MFCD01568483
CAS Number: 339106-98-8

Order Now

Specification Test Results

Melting point 239-240°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR