2-[(4-Chlorophenyl)sulfanyl]-1-methyl-1H-indole-3-carbaldehyde ≥95%
Supplier: Matrix Scientific
044165-500MG
044165-1G
044165-5G
101864-486EA
747.69
USD
101864-486
101864-488
101864-490
2-[(4-Chlorophenyl)sulfanyl]-1-methyl-1H-indole-3-carbaldehyde ≥95%
2-[(4-Chlorophenyl)sulfanyl]-1-methyl-1H-indole-3-carbaldehyde
Hazard: Irritant
Formula:
C₁₆H₁₂ClNOS Melting Pt: 154…156 °C |
MDL Number:
MFCD01315811 CAS Number: 338416-17-4 |
Specification Test Results
Melting point | 154-156°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...