2-Amino-4,5-dimethyl-1H-pyrrole-3-carbonitrile ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Amino-3-cyano-4,5-dimethylpyrrole

043941-500MG 043941-1G 043941-5G
101863-308EA 181.54 USD
101863-308 101863-310 101863-312
2-Amino-4,5-dimethyl-1H-pyrrole-3-carbonitrile ≥95%
2-Amino-4,5-dimethyl-1H-pyrrole-3-carbonitrile

Hazard: Irritant

Formula: C₇H₉N₃
Melting Pt: 164…166 °C
MDL Number: MFCD00849263
CAS Number: 21392-51-8

Order Now

Specification Test Results

Melting point 164-166°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR