4'-Methoxy-2'-nitroacetanilide ≥95%
Supplier: Matrix Scientific
Synonyms:
4-Acetamido-3-Nitroanisole, N-(4-Methoxy-2-nitro-phenyl)-acetamide, 4-Methoxy-2-nitroacetanilide, N-(4-Methoxy-2-nitrophenyl)acetamide
043515-500MG
043515-1G
043515-5G
101861-206EA
181.54
USD
101861-206
101861-208
101861-210
4'-Methoxy-2'-nitroacetanilide ≥95%
4'-Methoxy-2'-nitroacetanilide
Hazard: Irritant
Formula:
C₉H₁₀N₂O₄ MW: 210.19 g/mol Melting Pt: 117…118 °C |
MDL Number:
MFCD00017018 CAS Number: 119-81-3 |
Specification Test Results
Melting point | 117-118°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...