2-Amino-1-benzyl-4,5-dimethyl-1H-pyrrole-3-carbonitrile ≥95%
Supplier: Matrix Scientific
042688-500MG
042688-1G
042688-5G
101857-012EA
72.31
USD
101857-012
101857-014
101857-016
2-Amino-1-benzyl-4,5-dimethyl-1H-pyrrole-3-carbonitrile ≥95%
2-Amino-1-benzyl-4,5-dimethyl-1H-pyrrole-3-carbonitrile
Hazard: Irritant
Formula:
C₁₄H₁₅N₃ Melting Pt: 152…155 °C |
MDL Number:
MFCD01935932 CAS Number: 55817-72-6 |
Specification Test Results
Melting point | 152-155°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...