2-(Dicyclohexylphosphino)-2'-(dimethylamino)biphenyl ≥98%

Supplier: Matrix Scientific

Synonyms: DavePhos2'-(Dicyclohexylphosphino)-N,N-dimethyl-[1,1'-biphenyl]-2-amine2-Dicyclohexylphosphino-2’-(N,N-dimethylamino)biphenyl2-(Dicyclohexylphosphino)-2'-(N,N-dimethylamino))-1,1'-biphenyl

007179-1G
101792-142EA 44.62 USD
101792-142
2-(Dicyclohexylphosphino)-2'-(dimethylamino)biphenyl ≥98%
2-(Dicyclohexylphosphino)-2'-(dimethylamino)biphenyl

Hazard: Air Sensitive

Formula: C₂₆H₃₆NP
MW: 393.55 g/mol
Melting Pt: 115…119 °C
MDL Number: MFCD02183572
CAS Number: 213697-53-1

Order Now

Specification Test Results

Melting point 115-119°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR