2-(4-Methylphenyl)malonaldehyde
Supplier: Matrix Scientific
006851-1G
006851-5G
101791-326EA
387.69
USD
101791-326
101791-328
2-(4-Methylphenyl)malonaldehyde
2-(4-Methylphenyl)malonaldehyde
Hazard: Irritant
Formula:
C₁₀H₁₀O₂ Melting Pt: 131…134 °C |
MDL Number:
MFCD00216506 CAS Number: 27956-35-0 |
Specification Test Results
Melting point | 131-134°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...