2-Chloro-6-(trifluoromethyl)nicotinamide ≥98%

Supplier: Matrix Scientific

006800-250MG 006800-1G
101791-144EA 60 USD
101791-144 101791-146
2-Chloro-6-(trifluoromethyl)nicotinamide ≥98%
2-Chloro-6-(trifluoromethyl)nicotinamide

Hazard: Irritant

Formula: C₇H₄ClF₃N₂O
Melting Pt: 198…200 °C
MDL Number: MFCD01862659
CAS Number: 386704-05-8

Order Now

Specification Test Results

Melting point 198-200°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR