(2,3-Difluorophenyl)acetic acid ≥98%

Supplier: Matrix Scientific

Synonyms: 2-(2,3-Difluorophenyl)acetic acid

005577-1G 005577-5G 005577-25G
101789-162EA 15.38 USD
101789-162 101789-164 101789-166
(2,3-Difluorophenyl)acetic acid ≥98%
(2,3-Difluorophenyl)acetic acid

Hazard: Irritant

Formula: (C₆H₃F₂)CH₂COOH
Melting Pt: 118…119 °C
MDL Number: MFCD00040968
CAS Number: 145689-41-4

Order Now

Specification Test Results

Melting point 118-119°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR