N-Benzyl-2-(trifluoromethyl)pyridin-4-amine ≥95%

Supplier: Matrix Scientific

059631-500MG 059631-1G 059631-5G
101918-678EA 803.08 USD
101918-678 101918-680 101918-682
N-Benzyl-2-(trifluoromethyl)pyridin-4-amine ≥95%
N-Benzyl-2-(trifluoromethyl)pyridin-4-amine

Hazard: Irritant

Formula: C₁₃H₁₁F₃N₂
Melting Pt: 104…106 °C
MDL Number: MFCD18803520
CAS Number: 1280787-28-1

Order Now

Specification Test Results

Melting point 104-106°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR