7-Bromo-1-benzothiophene-2-carboxylic acid ≥95%

Supplier: Matrix Scientific

059546-500MG 059546-1G
101918-384EA 261.54 USD
101918-384 101918-386
7-Bromo-1-benzothiophene-2-carboxylic acid ≥95%
7-Bromo-1-benzothiophene-2-carboxylic acid

Hazard: Irritant

Formula: C₉H₅BrO₂S
Melting Pt: >275 °C
MDL Number: MFCD12032270
CAS Number: 19075-59-3

Order Now

Specification Test Results

Melting point 275°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR