(3R,4R)-1-Benzyl-4-(trifluoromethyl)tetrahydro-1H-pyrrole-3-carboxylic acid ≥95%

Supplier: Matrix Scientific

Synonyms: (3R,4R)-1-Benzyl-4-(trifluoromethyl)pyrrolidine-3-carboxylic acid

054686-500MG 054686-1G
101908-128EA 921.54 USD
101908-128 101908-130
(3R,4R)-1-Benzyl-4-(trifluoromethyl)tetrahydro-1H-pyrrole-3-carboxylic acid ≥95%
(3R,4R)-1-Benzyl-4-(trifluoromethyl)tetrahydro-1H-pyrrole-3-carboxylic acid

Hazard: Irritant

Formula: C₁₃H₁₄F₃NO₂
Melting Pt: 92 °C (dec.)
MDL Number: MFCD12974801
CAS Number: 184844-96-0

Order Now

Specification Test Results

Melting point 92°C(dec)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR