1-(4-Fluorobenzyl)-5-methyl-1H-1,2,3-triazole-4-carboxylic acid ≥95%

Supplier: Matrix Scientific

047467-500MG 047467-5G
101880-144EA 375.38 USD
101880-144 101880-148
1-(4-Fluorobenzyl)-5-methyl-1H-1,2,3-triazole-4-carboxylic acid ≥95%
1-(4-Fluorobenzyl)-5-methyl-1H-1,2,3-triazole-4-carboxylic acid

Hazard: Irritant

Formula: C₁₁H₁₀FN₃O₂
Melting Pt: 203 °C
MDL Number: MFCD06797392
CAS Number: 885950-27-6

Order Now

Specification Test Results

Melting point 203°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR