6,7-Dichloro-1,4-dihydro-2,3-quinoxalinedione2-oxime ≥95%

Supplier: Matrix Scientific

047147-500MG 047147-1G 047147-5G
101878-658EA 181.54 USD
101878-658 101878-660 101878-662
6,7-Dichloro-1,4-dihydro-2,3-quinoxalinedione2-oxime ≥95%
6,7-Dichloro-1,4-dihydro-2,3-quinoxalinedione2-oxime

Hazard: Irritant

Formula: C₈H₅Cl₂N₃O₂
Melting Pt: >300 °C
MDL Number: MFCD08689792
CAS Number: 177944-61-5

Order Now

Specification Test Results

Melting point 300°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR