Ethyl-4-(2-methoxyphenyl)-1,6-dimethyl-2-oxo-1,2,3,4-tetrahydro-5-pyrimidinecarboxylate ≥95%
Supplier: Matrix Scientific
047138-500MG
047138-1G
047138-5G
101878-606EA
181.54
USD
101878-606
101878-608
101878-610
Ethyl-4-(2-methoxyphenyl)-1,6-dimethyl-2-oxo-1,2,3,4-tetrahydro-5-pyrimidinecarboxylate ≥95%
Ethyl-4-(2-methoxyphenyl)-1,6-dimethyl-2-oxo-1,2,3,4-tetrahydro-5-pyrimidinecarboxylate
Hazard: Irritant
Formula:
C₁₆H₂₀N₂O₄ Melting Pt: 141…143 °C |
MDL Number:
MFCD00718193 CAS Number: 301321-51-7 |
Specification Test Results
Melting point | 141-143°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...