4-Benzoyldihydro-2(3H)-furanone ≥95%
Supplier: Matrix Scientific
047042-500MG
047042-1G
047042-5G
101878-168EA
181.54
USD
101878-168
101878-170
101878-172
4-Benzoyldihydro-2(3H)-furanone ≥95%
4-Benzoyldihydro-2(3H)-furanone
Hazard: Irritant
Formula:
C₁₁H₁₀O₃ Melting Pt: 62…64 °C |
MDL Number:
MFCD00463834 CAS Number: 21034-22-0 |
Specification Test Results
Melting point | 62-64°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...