1,4-Dihydro-2,3-quinoxalinedione-2-oxime ≥95%

Supplier: Matrix Scientific

047038-500MG 047038-1G 047038-5G
101878-156EA 181.54 USD
101878-156 101878-158 101878-160
1,4-Dihydro-2,3-quinoxalinedione-2-oxime ≥95%
1,4-Dihydro-2,3-quinoxalinedione-2-oxime

Hazard: Irritant

Formula: C₈H₇N₃O₂
Melting Pt: 262…264 °C
MDL Number: MFCD08689712
CAS Number: 177944-77-3

Order Now

Specification Test Results

Melting point 262-264°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR