3-Bromo-2-methyl-6-nitro-2H-indazole ≥95%

Supplier: Matrix Scientific

046979-500MG 046979-1G
101877-890EA 1109.23 USD
101877-890 101877-892
3-Bromo-2-methyl-6-nitro-2H-indazole ≥95%
3-Bromo-2-methyl-6-nitro-2H-indazole

Hazard: Irritant

Formula: C₈H₆BrN₃O₂
Melting Pt: 174…176 °C
MDL Number: MFCD02666041
CAS Number: 74209-41-9

Order Now

Specification Test Results

Melting point 174-176°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR