Ethyl-2-(3,4-dimethoxyphenyl)-4-hydroxy-1,3-thiazole-5-carboxylate ≥95%

Supplier: Matrix Scientific

046697-500MG 046697-1G 046697-5G
101876-598EA 278.46 USD
101876-598 101876-600 101876-602
Ethyl-2-(3,4-dimethoxyphenyl)-4-hydroxy-1,3-thiazole-5-carboxylate ≥95%
Ethyl-2-(3,4-dimethoxyphenyl)-4-hydroxy-1,3-thiazole-5-carboxylate

Hazard: Irritant

Formula: C₁₄H₁₅NO₅S
Melting Pt: 144…146 °C
MDL Number: MFCD08444004
CAS Number: 924868-99-5

Order Now

Specification Test Results

Melting point 144-146°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR