5-Iodo-2,4-dimethoxypyrimidine ≥98%

Supplier: Matrix Scientific

021424-1G 021424-5G
101812-064EA 40 USD
101812-064 101812-066
5-Iodo-2,4-dimethoxypyrimidine ≥98%
5-Iodo-2,4-dimethoxypyrimidine

Hazard: Irritant

Formula: C₆H₇IN₂O₂
Melting Pt: 71…75 °C
MDL Number: MFCD00090865
CAS Number: 52522-99-3

Order Now

Specification Test Results

Melting point 71-75°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR