2-Chloro-5-phenylnicotinonitrile ≥95%

Supplier: Matrix Scientific

044794-500MG 044794-1G 044794-5G
101867-632EA 307.69 USD
101867-632 101867-634 101867-636
2-Chloro-5-phenylnicotinonitrile ≥95%
2-Chloro-5-phenylnicotinonitrile

Hazard: Irritant

Formula: C₁₂H₇ClN₂
Melting Pt: 143…147 °C
MDL Number: MFCD00231610
CAS Number: 10177-10-3

Order Now

Specification Test Results

Melting point 143-147°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR