N-(2-Methoxyethyl)urea ≥95%

Supplier: Matrix Scientific

044550-500MG 044550-1G 044550-5G
101866-424EA 78.46 USD
101866-424 101866-426 101866-428
N-(2-Methoxyethyl)urea ≥95%
N-(2-Methoxyethyl)urea

Hazard: Irritant

Formula: C₄H₁₀N₂O₂
Melting Pt: 73…75 °C
MDL Number: MFCD03425768
CAS Number: 22033-10-9

Order Now

Specification Test Results

Melting point 73-75°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR