HyClone® Bovine Growth Serum, U.S. Origin
Supplier: Cytiva
SH30541.02 SH30541.03HI SH30541.03 SH30541.03IR25-40 SH30541.02HI
82006-906EA 62.7 USD
82006-906 82013-586 82006-908 76324-910 76635-868
HyClone® Bovine Growth Serum, U.S. Origin
Sera (Serum)
HyClone™ Bovine Growth Serum is a foetal bovine serum (FBS) replacement, consisting of bovine calf serum supplemented with chemically-defined components.
  • Sourced in the USA with complete traceability back to original source
  • Cost-effective alternative to FBS
  • Growth promotion studies demonstrate that BGS is an excellent replacement for FBS
  • Low in antibodies and high in growth factors
  • Virus panel testing according to 9 CFR 113.53
HyClone Bovine Growth Serum, U.S. Origin is supplemented with chemically defined components including amino acids, trace metals, vitamins, and other small molecules. Save money while ensuring excellent culture results with this cost-effective alternative to FBS. Bovine Growth Serum was developed for cell culturists who are concerned about the availability of fetal bovine serum and are seeking an affordable and viable alternative. BGS starts with Defined Bovine Calf Serum and is supplemented to stimulate cell growth and proliferation.
Origin is supplemented with chemically defined components including amino acids, trace metals, vitamins, and other small molecules.
Additional treatments available including heat-inactivation and irradiation. Additional testing, including EMEA-conforming testing, is also available.