Palladium(II) chloride ≥99%
Supplier: Matrix Scientific
|
Warning
|
Synonyms:
Palladium dichloride
069624-25G
069624-5G
101941-142EA
4600
USD
101941-142
101941-140
Palladium(II) chloride ≥99%
Palladium (II) chloride
Hazard code(s): Irritant
Formula:
PdCl₂ MW: 177.33 g/mol Melting Pt: 678 °C Density: 4 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003558 CAS Number: 7647-10-1 EINECS: 231-596-2 UN: 3260 ADR: 8,III Merck Index: 12,07122 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...