Methyl-10-undecenoate ≥95%

Supplier: Matrix Scientific

069103-1G 069103-10G
101938-944EA 747.69 USD
101938-944 101938-946
Methyl-10-undecenoate ≥95%
Methyl 10-undecenoate

Hazard code(s): Irritant

Formula: C₁₂H₂₂O₂
Boiling Pt: 248 °C (1013 hPa)
Melting Pt: –27.5 °C
Density: 0.889 g/cm³ (20 °C)
MDL Number: MFCD00016689
CAS Number: 111-81-9
EINECS: 203-910-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR