5-Aminosalicylic acid ≥97%

Supplier: Matrix Scientific

058320-100G 058320-500G
101914-580EA 63.08 USD
101914-580 101914-582
5-Aminosalicylic acid ≥97%
5-Aminosalicylic acid

Hazard code(s): Irritant

Formula: C₇H₇NO₃
Boiling Pt: 347 °C (1013 hPa)
Melting Pt: 280 °C (dec.)
Density: 1.568 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00007877
CAS Number: 89-57-6
EINECS: 201-919-1
Merck Index: 12,05964

Order Now

Specification Test Results

Melting point 280(dec.)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR