Pentafluorobenzyl chloride ≥98%

Supplier: Matrix Scientific

Synonyms: 1-(Chlorodifluoromethyl)-2,3,4-trifluorobenzeneα-Chloro-2,3,4,5,6-pentafluorotoluene2,3,4,5,6-Pentafluorobenzyl chloride

056581-1G
101910-948EA 84.62 USD
101910-948
Pentafluorobenzyl chloride ≥98%
Pentafluorobenzyl chloride

Hazard code(s): Irritant

Formula: (C₆F₅)CH₂Cl
MW: 216.54 g/mol
Boiling Pt: 72 °C (22mm)
Density: 1.59 g/cm³
MDL Number: MFCD17168377
CAS Number: 653-35-0
EINECS: 211-501-0
UN: 2922
ADR: 8,II

Order Now

Specification Test Results

Boiling point 72°C/22mm
Density 1.59

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR