2-Amino-3-iodopyridine ≥97% for synthesis

Supplier: Matrix Scientific

048724-1G 048724-5G
101885-020EA 121.54 USD
101885-020 101885-022
2-Amino-3-iodopyridine ≥97% for synthesis
2-Amino-3-iodopyridine

Hazard code(s): Irritant

Formula: C₅H₅IN₂
Melting Pt: 88…92 °C
MDL Number: MFCD01651885
CAS Number: 104830-06-0

Order Now

Specification Test Results

Melting point 88-92°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR