L(+)-Arginine
Supplier: Matrix Scientific
|
Warning
|
Synonyms:
H-L-Arg-OH, (S)-2-Amino-5-guanidinopentanoic acid, Arginine, (S)-(+)-Arginine, L-Arginine, H-Arg-OH
041282-25G
041282-100G
101851-186EA
21.54
USD
101851-186
101851-188
L(+)-Arginine
L(+)-Arginine
Formula:
C₆H₁₄N₄O₂ MW: 174.2 g/mol Boiling Pt: 368 °C (1013 hPa) Melting Pt: 238 °C Density: 1.3 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00002635 CAS Number: 74-79-3 EINECS: 200-811-1 Merck Index: 13,00785 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...