Adenine for synthesis

Supplier: Matrix Scientific
Danger

Synonyms: Purin-6-amine, 7H-Purin-6-amine, 6-Aminopurine

037413-25G 037413-100G 037413-500G
101843-316EA 23.08 USD
101843-316 101843-318 101843-320
Adenine for synthesis
Adenine
Formula: C₅H₅N₅
MW: 135.13 g/mol
Density: 0.99741 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00041790
CAS Number: 73-24-5
EINECS: 200-796-1
UN: 2811
ADR: 6.1 III
Merck Index: 13,00152

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR