1-Naphthylacetic acid
Supplier: Matrix Scientific
|
Warning
|
027536-250G
027536-1KG
101824-368EA
129.23
USD
101824-368
101824-370
1-Naphthylacetic acid
1-Naphthylacetic acid
Formula:
C₁₂H₁₀O₂ Boiling Pt: 352 °C (1013 hPa) Melting Pt: 129…131 °C Density: ∼1.263 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00004046 CAS Number: 86-87-3 EINECS: 201-705-8 Merck Index: 12,06458 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...