3-Amino-5-tert-butylisoxazole ≥97% for synthesis

Supplier: Matrix Scientific

021406-5G 021406-25G
101811-996EA 29.23 USD
101811-996 101811-998
3-Amino-5-tert-butylisoxazole ≥97% for synthesis
3-Amino-5-tert-butylisoxazole

Hazard code(s): Irritant

Formula: C₇H₁₂N₂O
Melting Pt: 110…111 °C
MDL Number: MFCD00055620
CAS Number: 55809-36-4

Order Now

Specification Test Results

Melting point 110-111°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR