2-Chlorobenzenesulfonamide

Supplier: Matrix Scientific

021039-25G 021039-100G
101811-234EA 21.54 USD
101811-234 101811-236
2-Chlorobenzenesulfonamide
2-Chlorobenzenesulphonamide

Hazard code(s): Irritant

Formula: C₆H₆ClNO₂S
Melting Pt: 189…191 °C
MDL Number: MFCD00051974
CAS Number: 6961-82-6
EINECS: 230-156-7

Order Now

Specification Test Results

Melting point 189-191°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR