4-Bromo-2-chlorobenzonitrile ≥97% for synthesis

Supplier: Matrix Scientific

020603-5G 020603-25G
101810-414EA 29.23 USD
101810-414 101810-416
4-Bromo-2-chlorobenzonitrile ≥97% for synthesis
4-Bromo-2-chlorobenzonitrile

Hazard code(s): Irritant

Formula: C₇H₃BrClN
Boiling Pt: 142…143 °C
Melting Pt: 71…73 °C
MDL Number: MFCD00040883
CAS Number: 154607-01-9
UN: 3439
ADR: 6.1,III

Order Now

Specification Test Results

Boiling point 142-143°C
Melting point 71-73°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR