3-Amino-5-bromopyridine ≥97% for synthesis

Supplier: Matrix Scientific
Warning

Synonyms: 5-Bromopyridin-3-amine, 5-Bromo-3-pyridylamine

020570-25G 020570-100G
101810-312EA 90.77 USD
101810-312 102832-898
3-Amino-5-bromopyridine ≥97% for synthesis
3-Amino-5-bromopyridine

Hazard code(s): Irritant

Formula: C₅H₅BrN₂
MW: 173.01
MDL Number: MFCD01646060
CAS Number: 13535-01-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR