2-Bromo-5-methylpyridine ≥98%

Supplier: Matrix Scientific
Warning

012520-25G 012520-100G
101802-746EA 46.15 USD
101802-746 101802-748
2-Bromo-5-methylpyridine ≥98%
2-Bromo-5-methylpyridine

Hazard code(s): Irritant

Formula: C₆H₆BrN
Boiling Pt: 95…96 °C (12mm)
Melting Pt: 41…43 °C
MDL Number: MFCD00209553
CAS Number: 3510-66-5

Order Now

Specification Test Results

Boiling point 95-96°C/12m
Melting point 41-43°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR