5-Bromo-2-fluoropyridine 98 for synthesis
Supplier: Matrix Scientific
011165-25G
011165-100G
101800-038EA
36.92
USD
101800-038
101800-040
5-Bromo-2-fluoropyridine 98 for synthesis
5-Bromo-2-fluoropyridine
Hazard code(s): Irritant
Formula:
C₅H₃BrFN MW: 175.99 g/mol Boiling Pt: 63 Density: 1.710 g/cm³ |
MDL Number:
MFCD01863742 CAS Number: 766-11-0 UN: 2810 ADR: 6.1 III |
Specification Test Results
Boiling point | 162-164°C/7 |
Density | 1.71 |
ND20 | 1.533 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...