6-Bromopicolinic acid ≥97% for synthesis

Supplier: Matrix Scientific

Synonyms: 6-Bromo-2-pyridinecarboxylic acid, 6-Bromopyridine-2-carboxylic acid

011076-25G 011076-100G
101799-784EA 89.23 USD
101799-784 101799-786
6-Bromopicolinic acid ≥97% for synthesis
6-Bromopicolinic acid

Hazard code(s): Irritant

Formula: C₆H₄BrNO₂
MW: 202.01
Melting Pt: 192…194 °C
MDL Number: MFCD00093197
CAS Number: 21190-87-4

Order Now

Specification Test Results

Melting point 192-194°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR