2-Amino-1,3,4-thiadiazole ≥97% for synthesis

Supplier: Matrix Scientific

Synonyms: [1,3,4]Thiadiazol-2-ylamine, 1,3,4-Thiadiazol-2-amine

010951-5G 010951-25G 010951-100G
101799-534EA 20 USD
101799-534 101799-536 101799-538
2-Amino-1,3,4-thiadiazole ≥97% for synthesis
2-Amino-1,3,4-thiadiazole

Hazard code(s): Irritant

Formula: C₂H₃N₃S
MW: 101.13
MDL Number: MFCD00003107
CAS Number: 4005-51-0
EINECS: 223-657-7
UN: 2811
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR