Tetrafluorophthalic anhydride ≥97%

Supplier: Matrix Scientific

009165-2.5G 009165-10G
101796-546EA 467.69 USD
101796-546 101796-548
Tetrafluorophthalic anhydride ≥97%
Tetrafluorophthalic anhydride

Hazard code(s): Irritant, Moisture Sensitive

Formula: C₈F₄O₃
MW: 220.08 g/mol
Melting Pt: 94…96 °C
MDL Number: MFCD00039697
CAS Number: 652-12-0
EINECS: 211-485-5

Order Now

Specification Test Results

Melting point 94-96°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR