3-Iodobenzoic acid ≥98% for synthesis
Supplier: Matrix Scientific
Synonyms:
m-Iodobenzoic acid
008699-25G
008699-100G
101795-208EA
21.54
USD
101795-208
101795-210
3-Iodobenzoic acid ≥98% for synthesis
3-Iodobenzoic acid
Hazard code(s): Irritant, Light Sensitive
Formula:
C₇H₅IO₂ Boiling Pt: 337 °C (1013 hPa) Melting Pt: 186…188 °C Density: 2.171 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00002496 CAS Number: 618-51-9 EINECS: 210-555-2 |
Specification Test Results
Melting point | 186-188°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...