2-Aminoquinoline ≥98%

Supplier: Matrix Scientific

Synonyms: Quinolin-2-ylamine

007505-1G 007505-5G
101792-942EA 69.23 USD
101792-942 101792-944
2-Aminoquinoline ≥98%
2-Aminoquinoline

Hazard code(s): Irritant, Irritant-Harmful, Air Sensitive

Formula: C₉H₈N₂
MW: 144.18 g/mol
Boiling Pt: 305 °C (1013 hPa)
Melting Pt: 129 °C
MDL Number: MFCD00038079
CAS Number: 580-22-3
EINECS: 209-458-8

Order Now

Specification Test Results

Melting point 129°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR