1-(Difluoromethoxy)-3-nitrobenzene ≥98%

Supplier: Matrix Scientific

006014-5G 006014-25G
101789-914EA 178.46 USD
101789-914 101789-916
1-(Difluoromethoxy)-3-nitrobenzene ≥98%
1-(Difluoromethoxy)-3-nitrobenzene

Hazard code(s): Irritant

Formula: C₇H₅F₂NO₃
MDL Number: MFCD03407974
CAS Number: 22236-07-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR