Pentafluorobenzoic acid ≥99%

Supplier: Matrix Scientific
Warning

005946-25G 005946-100G 005946-500G
101789-822EA 47.69 USD
101789-822 101789-824 101789-826
Pentafluorobenzoic acid ≥99%
Pentafluorobenzoic acid

Hazard code(s): Irritant

Formula: C₇HF₅O₂
Boiling Pt: 220 °C
Melting Pt: 100…102 °C
Density: 1.942 g/cm³ (22 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002406
CAS Number: 602-94-8
EINECS: 210-026-6

Order Now

Specification Test Results

Boiling point 220°C
Melting point 100-102°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR