3-(Trifluoromethyl)cinnamic acid ≥98% for synthesis

Supplier: Matrix Scientific
Warning

Synonyms: 3-(3-(Trifluoromethyl)phenyl)acrylic acid

005397-5G 005397-25G
101788-842EA 29.23 USD
101788-842 101788-844
3-(Trifluoromethyl)cinnamic acid ≥98% for synthesis
3-(Trifluoromethyl)cinnamic acid

Hazard code(s): Irritant

Formula: C₁₀H₇F₃O₂
MW: 216.16 g/mol
Boiling Pt: 292
Melting Pt: 135…137 °C
MDL Number: MFCD00004393
CAS Number: 779-89-5
EINECS: 212-301-6

Order Now

Specification Test Results

Melting point 135-137°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR