2-(Trifluoromethoxy)benzyl alcohol ≥97% for synthesis

Supplier: Matrix Scientific

005378-1G 005378-5G
101788-784EA 29.23 USD
101788-784 101788-786
2-(Trifluoromethoxy)benzyl alcohol ≥97% for synthesis
2-(Trifluoromethoxy)benzyl alcohol

Hazard code(s): Irritant

Formula: C₈H₇F₃O₂
MW: 192.14
Boiling Pt: 96…98 °C (12mm)
MDL Number: MFCD00153285
CAS Number: 175278-07-6

Order Now

Specification Test Results

Boiling point 96-98°C/12m


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR