1-Iodo-4-(trifluoromethoxy)benzene ≥97% for synthesis

Supplier: Matrix Scientific

Synonyms: 4-(Trifluoromethoxy)iodobenzene

005359-25G 005359-100G
101788-746EA 21.54 USD
101788-746 101788-748
1-Iodo-4-(trifluoromethoxy)benzene ≥97% for synthesis
1-Iodo-4-(trifluoromethoxy)benzene

Hazard code(s): Irritant, Light Sensitive

Formula: C₇H₄F₃IO
Boiling Pt: 72…74 °C (15mm)
Density: 1.909 g/cm³
MDL Number: MFCD00042411
CAS Number: 103962-05-6

Order Now

Specification Test Results

Boiling point 72-74°C/15m


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR