1,3-Dichloro-4-fluorobenzene ≥99% for synthesis

Supplier: Matrix Scientific
Warning

Synonyms: 2,4-Dichlorofluorobenzene, 2,4-Dichloro-1-fluorobenzene

005278-25G 005278-100G
101788-570EA 18.46 USD
101788-570 101788-572
1,3-Dichloro-4-fluorobenzene ≥99% for synthesis
1,3-Dichloro-4-fluorobenzene

Hazard code(s): Irritant

Formula: C₆H₃Cl₂F
Boiling Pt: 172…174 °C
Melting Pt: –23 °C
Density: 1.409 g/cm³
MDL Number: MFCD00000580
CAS Number: 1435-48-9
EINECS: 406-160-1
UN: 2810
ADR: 6.1,III

Order Now

Specification Test Results

Boiling point 172-174°C
Density 1.409
ND20 1.525


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR