1,6-Diiodoperfluorohexane ≥97% for synthesis

Supplier: Matrix Scientific

Synonyms: Perfluoro-1,6-diiodohexane

004921-5G 004921-25G
101787-994EA 444.62 USD
101787-994 101787-996
1,6-Diiodoperfluorohexane ≥97% for synthesis
1,6-Diiodoperfluorohexane

Hazard code(s): Irritant, Light Sensitive, Refrigerate

Formula: C₆F₁₂I₂
Boiling Pt: 115 °C (100mm)
Melting Pt: 25 °C
Density: 2.357 g/cm³
MDL Number: MFCD00042264
CAS Number: 375-80-4
EINECS: 206-794-7

Order Now

Specification Test Results

Boiling point 115°C/100mm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR