4-Iodo-3-nitroanisole ≥97% for synthesis

Supplier: Matrix Scientific
Warning

Synonyms: 1-Iodo-4-methoxy-2-nitrobenzene

004652-5G 004652-25G
101787-622EA 24.62 USD
101787-622 101787-624
4-Iodo-3-nitroanisole ≥97% for synthesis
4-Iodo-3-nitroanisole

Hazard code(s): Irritant, Light Sensitive

Formula: C₇H₆INO₃
MW: 279.03 g/mol
Melting Pt: 61…62 °C
MDL Number: MFCD00024195
CAS Number: 58755-70-7

Order Now

Specification Test Results

Melting point 61-62°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR